Learn More
Medchemexpress LLC 1-Pyrrolidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, (3R)- | 147081-49-0 | MFCD03419272 | 100.0% | 186.25 | 10 G
Supplier: Medchemexpress LLC HY4009710G
(R)-1-Boc-3-aminopyrrolidine is a biochemical reagent and organic compound used in life science research. It also serves as a chiral building block for synthesizing pharmaceuticals and agrochemicals.
- Biochemical reagent
- Biological material
- Organic compound
- Used in life science research
- Chiral building block for synthesis