Learn More
Medchemexpress LLC 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine | 59403-52-0 | MFCD00214381 | 98.0% | 834.2 g/mol | C48H84NO8P | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW4409831MG
Encompass_Preferred
SDPC (1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine) is a docosahexaenoic acid (DHA)-containing phosphatidylcholine used in lipid biochemistry and membrane research. It supplies a DHA residue at the sn-2 position and is intended for preparing model membranes, lipidomics studies, and assays probing lipid-protein interactions and membrane properties.
  • High purity DHA-containing phospholipid suitable for biochemical studies.
  • Facilitates construction of DHA-enriched model membranes and liposomes.
  • Compatible with lipidomics and mass spectrometry workflows.
  • Useful for studying membrane fluidity, signaling, and lipid-protein interactions.
  • Available in multiple small pack sizes for analytical use.
Catalog No. 50-002-80674