Learn More
Medchemexpress LLC 1-stearoyl-2-palmitoyl-sn-glycero-3-phosphocholine | 59403-53-1 | MFCD00672603 | 100.0% | 762.09 g/mol | C42H84NO8P | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW44098110MG
Encompass_Preferred
SPPC (1-stearoyl-2-palmitoyl-sn-glycero-3-phosphocholine) is a glycerophosphocholine phospholipid used as a research reagent and liposome component. The sn-1 position contains stearic acid (18:0) and the sn-2 position contains palmitic acid (16:0). It is commonly employed in liposome formulation, membrane biophysics, and other in vitro membrane studies.
  • Provides defined 18:0/16:0 phosphatidylcholine composition.
  • Suitable for liposome formulation and membrane biophysical studies.
  • Reported high purity for reliable experimental results.
  • Soluble in ethanol (100 mg/mL) with ultrasonic assistance.
  • Powder stable at -20°C for long-term storage; in-solvent stability is shorter.
Catalog No. 50-002-81161