Learn More
Medchemexpress LLC 1-(tert-butoxycarbonyl)azetidine-3-carboxylic acid | 142253-55-2 | MFCD01860897 | 98.3% | 201.22 g/mol | C9H15NO4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY401411G
Encompass_Preferred
1-(tert-butoxycarbonyl)azetidine-3-carboxylic acid is a Boc-protected azetidine derivative used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain-based PROTAC linker. It is supplied as a high-purity white crystalline powder, suitable for medicinal chemistry and organic synthesis workflows.
  • Boc-protected azetidine scaffold suitable for linker chemistry.
  • High purity for reliable synthesis and characterization.
  • Available in multiple package sizes for small-scale and bulk use.
  • Soluble in DMSO for common synthetic procedures.
  • Systematic name and identifiers available for database referencing.
Catalog No. 50-002-02323