Learn More
Selleck Chemical LLC 1-Thioglycerol E2907-25MG
SDP
Supplier:  Selleck Chemical LLC E290725MG
Encompass_Preferred
1-Thioglycerol(Mercaptopropanedioltech) is an inhibitor of glycerol kinase activity It stimulates the synthesis of porphyrin in aerobically growing Escherichia coli
Catalog No. 50-000-40122