Learn More
Medchemexpress LLC 1-Tridecanol-d27 | 203633-18-5 | 227.53 | 25 MG
Supplier: Medchemexpress LLC 25MGHYW004293S
1-Tridecanol-d27 is a deuterium-labeled 1-Tridecanol. Stable heavy isotopes of hydrogen have been incorporated into drug molecules for various applications, largely as tracers for quantitation during the drug development process. Deuteration has also gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product is for research use only and has not been fully validated for medical applications.
- Isotope-labeled compound
- Suitable for research use only
- Deuterium-labeled for pharmacokinetic studies
- Potential to affect drug metabolism