Learn More
Medchemexpress LLC 1-Undecanol | 112-42-5 | 172.31 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00429210G
Encompass_Preferred
1-Undecanol (Undecyl alcohol) is a chemical compound primarily generated from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from the soil. It has been observed to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Primarily generated from the degradation of 2-tridecanone by Pseudomonas bacteria.
  • Enhances the attraction of Grapholita molesta to sex pheromone traps.
  • Acts as a synergist for Grapholita molesta sex pheromone.
Catalog No. 50-004-45893