Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Undecanol | 112-42-5 | MFCD00004751 | 172.31 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW004292

Encompass_Preferred

1-Undecanol, also known as Undecyl alcohol, is primarily generated from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from soil. This compound has been observed to enhance the attraction of Grapholita molesta to sex pheromone traps.

  • Purity: 99.8%
  • Enhances attraction of Grapholita molesta to sex pheromone traps
  • Appearance: solid below 11°C, liquid above 11°C
  • Color: colorless to light yellow
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-004-48268


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.