Learn More
Medchemexpress LLC 10-[[1-[(2,4-dichlorophenyl)methyl]indazole-3-carbonyl]amino]decyl-triphenylphosphanium;bromide | 2361564-49-8 | 97.0% | C43H45BrCl2N3OP | 10MG
SDP
Supplier:  Medchemexpress LLC HY13483210MG
Encompass_Preferred
Mito-LND (Mito-Lonidamine) is a mitochondria-targeted inhibitor of oxidative phosphorylation used in preclinical research to disrupt mitochondrial bioenergetics, elevate reactive oxygen species, and trigger autophagic cell death in cancer models. It is supplied as a solid or as DMSO solution with defined purity and storage recommendations for laboratory use.
  • Mitochondria-targeted OXPHOS inhibitor useful for studying mitochondrial metabolism.
  • Induces reactive oxygen species and autophagic cell death in tumor cells.
  • Available as solids and DMSO solutions in multiple pack sizes.
  • High purity suitable for research applications.
  • Stable under recommended sealed storage conditions at -20°C; longer-term stability in solvent at -80°C.
Catalog No. 50-002-58992