Learn More
Cayman Chemical 10 11-Cl-BBQ 5mg
SDP
Supplier:  Cayman Chemical 299785 MG
Encompass_Preferred
A mixture of 10-Cl-BBQ and 11-Cl-BBQ that acts as an AhR agonist; induces Cyp1a1 transcription in mouse liver at 10 mg/kg
Catalog No. 50-297-8737