Learn More
Cayman Chemical 10 11-EDP EthnolamIde 100ug
SDP
Supplier:  Cayman Chemical 25878100 UG
Encompass_Preferred
An ω-3 endocannabinoid epoxide and CB receptor agonist (EC50s = 0.43 and 22.5 nM for CB1 and CB2 receptors, respectively); reduces the viability of 143B metastatic osteosarcoma cells at 12.5 and 25 μM; induces apoptosis and inhibits cell migration in a wound-healing assay in 143B, MG63, and HOS osteosarcoma cells; reduces tube formation by HUVECs in a Matrigel™ assay
Catalog No. 50-293-5564