Learn More
Cayman Chemical 10 11dIhdro10hdrox Carb 10mg
SDP
Supplier:  Cayman Chemical 1846710 MG
Encompass_Preferred
An antiepileptic agent and active metabolite of oxcarbazepine; a racemic mixture of (R)- and (S)-10,11-dihydro-10-hydroxy carbamazepine
Catalog No. 50-296-6221