Learn More
Medchemexpress LLC 10,13-dimethyl-17-(trifluoromethylsulfonyloxy)-cyclopenta[a]phenanthren-3-yl acetate | 115375-60-5 | MFCD18252939 | 99.1% | 462.52 g/mol | C22H29F3O5S | 10 MG

Supplier: Medchemexpress LLC HY7562410MG
Synthetic steroid triflate acetate supplied for laboratory research as a solid reagent or intermediate in steroid chemistry. It is intended for analytical and synthetic applications and is not for human or veterinary use.
- Steroid triflate acetate structure suitable as a synthetic intermediate.
- Formula: C22H29F3O5S.
- Molecular weight: 462.52 g/mol.
- Purity: 99.09% (HPLC).
- Appearance: white to light yellow solid; typical pack size 10 mg.
- CAS number: 115375-60-5.
- Intended for research use only; handle per safety data sheet instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.