Learn More
J Kem Scientific Inc 10/18 TAPER
SDP
Supplier:  J Kem Scientific Inc 1018
Encompass
PTFE Probe Adapters, Probe Diameter: 1/8 in. , Joint Size: 10/18, PTFE probe adapters form an air and vacuum tight seal between thermocouple probes and standard taper joints
Catalog No. NC9731572