Learn More
Chemglass Life Sciences 10/30 Inner Joint, Un-Cut Drip, on 8mm OD x 1.5mm
SDP
Supplier:  Chemglass Life Sciences CG11705
Encompass_Preferred
Full or medium length standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints. 10/30 Inner Joint, Un-Cut Drip, on 8mm OD x 1.5mm.
Catalog No. CG11705