Learn More
Cayman Chemical 10-Acetyl-3,7-dihydroxyphenoxazine, 119171-73-2, 5 mg
SDP
Supplier:  Cayman Chemical 100104695
Encompass_Preferred
Molecular Formula C14H11NO4, Purity ≥98%, Formula Weight 257.2, Synonyms: 1-(3,7-dihydroxy-10H-phenoxazin-10-yl)-ethanone, A 6550, ADHP, Amplex Red; A highly sensitive, stable substrate for HRP that reacts with H2O2 to produce the fluorescent compound resorufin; enables detection of H2O2 at a concentration as low as 5 pmol per 100 l samples.
Catalog No. NC1337762