Learn More
Seracare Life Sciences Inc 10% BSA DILUENT/BLOCKING SOL
Supplier: Seracare Life Sciences Inc 51400006
Convenient antibody diluent/blocking solution concentrate containing 10% bovine serum albumin in phosphate buffer. Compatible with most assay formats.