Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-hydroxy-12(Z)-octadecenoic acid | 34932-12-2 | 99.8% | 298.46 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY165772

Encompass_Preferred

10-Hydroxy-12(Z)-octadecenoic acid is an ester product with a molecular weight of 298.46 and a chemical formula of C18H34O3. It is a solid, white to off-white compound with a purity of 99.82%, intended for research use only. For in vitro studies, it can be dissolved in DMSO up to 100 mg/mL. For in vivo experiments, it is formulated into clear solutions (≥ 2.5 mg/mL) using co-solvents like PEG300, Tween-80, Saline, 20% SBE-β-CD in Saline, or Corn Oil.

  • High purity: Exhibiting a purity of 99.82%.
  • Versatile solubility: Soluble in DMSO for in vitro studies and can be formulated with various co-solvents for in vivo applications, allowing for flexible experimental designs.
  • Solid form: Provided as a solid, white to off-white in color, facilitating handling and storage.
  • Research grade: Specifically designed for research purposes.

Catalog No. 50-003-92992


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.