Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-hydroxy-12(z)-octadecenoic acid | 34932-12-2 | 99.8% | 298.46 g/mol | C18H34O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16577210MG

Encompass_Preferred

10-Hydroxy-12(Z)-octadecenoic acid is a research-grade hydroxy monounsaturated fatty acid supplied as an ester product for laboratory use. It is intended for in vitro biochemical and cellular research and is provided with supporting analytical and safety documentation.

  • High purity: 99.82% reported.
  • Molecular weight: 298.46 g/mol.
  • Chemical formula: C18H34O3.
  • CAS number: 34932-12-2.
  • Supplied in small milligram pack sizes suitable for analytical and preparative work.
  • Certificate of analysis (COA) and safety data sheet (SDS) available.
  • Suitable as a bioactive lipid reagent for biochemical and cellular assays.

Catalog No. 50-002-89240


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.