Learn More
Enzo Life Sciences 10-Hydroxycamptothecin (20mg). CAS: 64439-81-2
SDP
Supplier:  Enzo Life Sciences BMLGR3160020
Encompass_Preferred
A DNA topoisomerase I inhibitor that induces differentiation in HL-60 cells. It induces apoptosis in human breast cancer cell lines MCF-7 and MDA-MB-468. Purity: ≥97%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0968