Learn More
Vortex Sales Group 10 Micron Direct Fit Dissolution Cannula Filters (Jar Of 1000)
Supplier:  Vortex Sales Group PSFIL010VK1000
Encompass_Preferred
10 Micron direct fit Dissolution Cannula Filters, made of UHMW Polyethylene, fits securely on the end of a Agilent/Varian cannula sample probe, Jar of 1000
Catalog No. NC1645649