Learn More
Perkin Elmer US LLC 1.0 mL Sample Loop, 1 mm ID, connect to ports #1 and #4
Supplier:  Perkin Elmer US LLC N0777664
Encompass_Preferred
Multipurpose PFA FAST loops for high-flow valves. Recommended for use with 1.0 mm i.d. (gray) probe. 1.6 mm o.d., 1/4-28 fittings.
Catalog No. 50-211-2152