Learn More
Bellco Glass, Inc. 10 mL Sterile Disposable Sampler Tubes with Caps on, Sterile, 50/pk, 1000/cs
SDP
Supplier:  Bellco Glass, Inc. 451400743
Encompass_Preferred
10 mL Sterile Disposable Sampler Shorter Tubes with Caps on, Sterile, 50/pk, 1000/cs. Transport tubes are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Used to store, transport or freeze samples. USP VI Polypropylene Tubes, HDPE Caps.Self-standing: the vial placed on the lab workbench steadily even without a base.Electron Beam Sterilization, SAL=10-6: safe, fast, environment-friendly, no chemical residues.100,000-level Clean Workshop Production, DNAse-free, RNAse-free, pyrogen-free, endotoxin-free.Temperature Range: Approx.-196°C to 121°CExcellent sealing performance, no leakage of negative pressure 95Kpa, ensuring safe and effective air transportation.RNase and pyrogen freeElectron beam sterilization, SAL=10-6
Catalog No. 50-233-0463