Learn More
Medchemexpress LLC 10-nitrooleic acid | 875685-46-4 | MFCD12912274 | 98.9% | 327.46 | C18H33NO4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1015591MG
Encompass_Preferred
10-nitrooleic acid (CXA-10) is an endogenous nitro fatty acid used in research as a modulator of redox-sensitive signaling pathways, including Nrf2 activation and NF-κB inhibition. It is investigated for effects on oxidative stress, inflammation, and fibrosis, and is supplied as a high-purity liquid for biochemical and cellular studies.
  • Modulates Nrf2 and NF-κB signaling, useful in redox biology research.
  • Investigated for anti-inflammatory and anti-fibrotic effects in disease models.
  • High purity (98.89% by LCMS) suitable for analytical and biological assays.
  • Provided as a liquid, colorless to light yellow, facilitating solution preparation.
  • Molecular formula C18H33NO4; molecular weight 327.46 g/mol for stoichiometric calculations.
  • Available in small-volume quantities for dose-response and pilot studies.
Catalog No. 50-002-47920