Learn More
Grainger End Cap: 10 Series, 1 in x 1/8 in x 1 in, Black, Dust Cover, Matte, Single, For 0.3 in Slot Width, 2 per Pack
Supplier:  Grainger 2RCR7
Encompass_Preferred
End caps cover the open ends on T-slot framing profiles to mask the sharp edges on the ends of the profiles and provide a finished look. They also prevent dust and dirt from entering the profiles, which helps keep them clean.
Catalog No. NC2885241