Learn More
Medchemexpress LLC [10]-Shogaol | 36752-54-2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN24345MG
Encompass_Preferred
[10]-Shogaol is an orally active antioxidant extracted from ginger (*Zingiber officinale*). It demonstrates various biological activities including anti-inflammatory and anticancer effects. It is suitable for laboratory research in immunology, inflammation, and anti-infection pathways.
  • Inhibits COX-2 and NF-κB phosphorylation
  • Reduces proinflammatory cytokines such as IL-1β and IL-6
  • Exhibits anti-inflammatory properties
  • Demonstrates anticancer effects against Docetaxel-resistant prostate cancer
  • Shows larvicidal activity against *Angiostrongylus cantonensis* L5 larvae
Catalog No. 50-004-20982