Learn More
Electron Microscopy Sciences 10 1/4" Specimen Gripper
SDP
Supplier:  Electron Microscopy Sciences 72928
Encompass_Preferred
Stainless steel, long, serrated fine tips, for easy removal of specimens from deep containers. Scissor type.
Catalog No. 50-288-10