Learn More
ATCC 10 strain even mix; genomic material
Supplier:  ATCC MSA1000
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises genomic DNA prepared from fully sequenced, characterized, and authenticated ATCC Genuine Cultures that were selected based on relevant phenotypic and genotypic attributes, such as Gram stain, GC content, genome size, and spore formation. NGS Standards enable the optimization of metagenomics workflows and research applications, providing reliable comparative data while improving assay consistency. Each order for this product is provided with access to run 20 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics.
Catalog No. 50-238-1947