Learn More
Pacer Global LLC 100mL Reservoir, White, Sterile, 5/pack, 200/case
SDP
Supplier:  Pacer Global LLC MLSMRWS100/DEL
Encompass_Preferred
100mL Disposable Reagent Reservoir. Sterile. Polystyrene. White. Made from virgin grade resin. Lot Certified RNase/DNase-free. Non-Pyrogenic, Animal Free, Human DNA Free. Designed to accommodate single & multichannel pipettors. Stacking lugs prevent crushing & facilitate separation. Graduation marks on the interior walls. Pour-off spouts in each corner. 5/pack, 200/case.
Catalog No. NC2065671
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product