Learn More
Chemglass Life Sciences Automation Filter Tips for Tecan Systems, Clear, 1000 μL, Sterile, Blister Packed (CLS-3499A-106)
SDP
Supplier:  Chemglass Life Sciences CLS3499A106
Encompass_Preferred
1000 μL automation filter pipette tips designed for high-throughput liquid handling on laboratory automation systems. Clear, sterile, blister-packed nest tips provide aerosol-resistant filtration and are manufactured from medical-grade polypropylene to minimize contamination risk in genomics, proteomics, and biopharmaceutical workflows.
  • 1000 μL capacity for large-volume transfers.
  • Designed for common high-throughput liquid-handling workstations.
  • Sterile; electron beam sterilized.
  • Medical-grade polypropylene; DNase/RNase free.
  • Aerosol-resistant filter to protect samples and instruments.
  • Packaging: 96 x 2 blister per pack; 2,304 per case.
  • Produced in a 100,008 grade clean room.
Catalog No. 50-273-3242