Learn More
Chemglass Life Sciences 1000 μL Automation Clear Filter Tips for Hamilton (CLS-3499A-046)
SDP
Supplier:  Chemglass Life Sciences CLS3499A046
Encompass_Preferred
Chemglass Life Sciences sterile 1000 μL clear filter tips designed for automated pipetting on Hamilton systems. Tips include an internal aerosol barrier to protect instruments and are supplied in nest-packed sterile units for high-throughput workflows.
  • Sterile, DNase/RNase free medical-grade polypropylene tips.
  • Integrated filter provides aerosol protection for pipetting systems.
  • Clear tips for easy sample visibility during transfer.
  • Packaged 96 tips per pack, 2,304 tips per case (24 packs per case).
  • Manufactured in a controlled clean room to minimize contamination risk.
Catalog No. 50-273-3265