Learn More
Chemglass Life Sciences Chemglass Automation Clear Filter Tips for Tecan, 1000 μl, Sterile, Boxed (96/rack)
SDP
Supplier:  Chemglass Life Sciences CLS3499A406
Encompass_Preferred
1000 μl automation clear filter tips compatible with high-throughput liquid handling workflows. Made from medical-grade polypropylene and produced in clean-room conditions to minimize contamination; aerosol-resistant filters protect samples and instruments.
  • 1000 μl capacity for large-volume transfers.
  • Sterile, electron beam sterilized.
  • Medical-grade polypropylene construction.
  • DNase/RNase free and produced in a clean-room environment.
  • Aerosol-resistant filter to protect samples and instruments.
  • Boxed packaging: 96 tips per rack, 10 racks per pack, 50 racks per case.
Catalog No. 50-273-3232