Learn More
Chemglass Life Sciences 1000 μL automation tips for Tecan systems, clear, sterile - 96 x 2 blister (2,304/case)
SDP
Supplier:  Chemglass Life Sciences CLS3499A105
Encompass_Preferred
Clear 1000 μL robotic tips designed for use with Tecan liquid-handling systems. Sterile and DNase-/RNase-free, these medical-grade polypropylene tips are supplied preloaded in blister packs for automated loading and consistent high-throughput performance.
  • Volume: 1000 μL per tip.
  • Sterile and DNase- and RNase-free.
  • Made from medical-grade polypropylene for chemical resistance and dimensional stability.
  • Preloaded in 96 x 2 blister packs for automated rack handling.
  • Case pack: 2,304 tips.
  • Electron-beam sterilized and produced in a high-grade cleanroom.
Catalog No. 50-273-3243