Learn More
Chemglass Life Sciences 1000 μL Automation clear tips for Hamilton systems, aerosol-resistant filter, sterile, 96/rack (10 racks/pk, 50 racks/cs)
SDP
Supplier:  Chemglass Life Sciences CLS3499A066
Encompass_Preferred
Clear 1000 μL automation pipette tips with aerosol-resistant filters, produced in a clean-room and supplied sterile for use with Hamilton automated liquid-handling systems. Packaged in nests with 96 tips per rack for high-throughput workflows.
  • Volume: 1000 μL.
  • Compatibility: automated liquid-handling systems.
  • Packaging: box-packed, nest; 96 tips per rack.
  • Pack configuration: 10 racks per pack, 50 racks per case.
  • Sterilization: electron beam sterilized.
  • Filter: aerosol-resistant filter included.
  • Material: medical-grade polypropylene.
  • Clean-room production: manufactured in a clean-room environment.
  • Free of DNase and RNase.
Catalog No. 50-273-3255