Learn More
Chemglass Life Sciences Automation Conductive Filter Pipette Tips, 1000 μL for Hamilton Automated Systems, Sterile, Barcode, 96/Pack
SDP
Supplier:  Chemglass Life Sciences CLS3499A013
Encompass_Preferred
Automation conductive filter pipette tips (1000 μL) designed for use with Hamilton automated liquid-handling systems. These conductive, aerosol-resistant filter tips provide contamination control and instrument protection during high-throughput automated pipetting. Sterile, DNase/RNase-free, and supplied in barcode-enabled blister nests for rack traceability and automation compatibility.
  • 1000 μL nominal volume
  • Conductive, aerosol-resistant filter
  • Sterile; electron beam sterilized
  • DNase/RNase free
  • Medical-grade polypropylene construction
  • Packaged 96 tips per pack; 2,304 tips per case
  • Barcode-enabled nest packaging for traceability
  • Manufactured in a 100,049-grade clean room
Catalog No. 50-273-3279