Learn More
Chemglass Life Sciences 1000mL Funnel Only, Coarse Frit, #50 O-Ring Joint, 90 O.D., 75 Pinchclamp
SDP
Supplier:  Chemglass Life Sciences CG140516
Encompass_Preferred
1000mL Funnel Only, Coarse Frit, #50 O-Ring Joint, 90 O.D., 75 Pinchclamp. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.
Catalog No. CG140516