Learn More
Chemglass Life Sciences Vacuum Trap, 1000mL Capacity, Fits 4300mL Dewar Flask, 400mm OAH x 130mm Width, Complete
SDP
Supplier:  Chemglass Life Sciences CG453102
Encompass_Preferred
Vacuum Trap, 1000mL Capacity, Fits 4300mL Dewar Flask, 400mm OAH x 130mm Width, Complete. Similar to CG-4529, but having a 40mm o-ring joint to connect the trap body to the stopper. O-ring joint provides for a vacuum-tight grease-free seal and permits the trap body to be easily disconnected from the stopper for cleaning. Complete trap is supplied with a stopper, trap body and CG-150-09 clamp.
Catalog No. CG453102