Learn More
SPEX CERTIPREP LLC 3P Fluoride from Sodium Fluoride at 1,000 ug/mL in H2O, 100 mL
Supplier:  SPEX CERTIPREP LLC 3P ICFFM100
Encompass_Preferred
(HPS by Zeptometrix) Fluoride Anion IC Single-Component Standard from Sodium Fluoride at 1,000 ug/mL in H2O.
Catalog No. NC1734466