Learn More
Accelagen 1000U HRV3C PROTEASE GST/HIS
Supplier:  Accelagen H0101S
Encompass
Turbo3C Protease is the most user-friendly HRV3C protease and offers the advantage of a dual His/GST tag. Turbo3C is a high specific restriction grade protease with robust activity and excellent stability. It recognizes the PreScission site of LEVLFQGP and cleaves between Q and G.
Catalog No. NC9694315