Learn More
CELLTREAT Scientific Products 1000L Extended Length Pipette Tips, Bulk Packed, Non-sterile
Supplier: CELLTREAT Scientific Products 229049
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Extended length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.