Learn More
CELLTREAT Scientific Products 1000µL Pipette Tip Reload System, LfTS Fit, Non-sterile
SDP
Supplier:  CELLTREAT Scientific Products 229088
Encompass_Preferred
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tipsLfTS Reload Systems offer a convenient and economical way to refill empty tip racks, saving both time and moneyReload Systems dramatically reduce plastic waste by reusing empty racks10μL extended length tips have a maximum volume of 20μL and are compatible with 2, 10, and 20μL pipettesFine tip for improved accuracy and precision
Catalog No. NC3371308