Learn More
Chemglass Life Sciences Chemglass 1000 μL Pipette Tips, Clear, Bulk Nest (1,000/pk, 10,000/cs)
SDP
Supplier:  Chemglass Life Sciences CLS3499E100
Encompass_Preferred
Chemglass 1000 μL clear pipette tips are non-sterile, bulk-packaged tips designed for general-purpose liquid handling and high-throughput workflows. Made from ultra-clear polymer, they provide visual clarity for accurate aspiration and are supplied in nest packaging for efficient bulk loading.
  • 1000 μL capacity.
  • Ultra-clear polymer construction.
  • Non-sterile, bulk nest packaging.
  • Supplied 1,000 units per pack and 10,000 units per case.
  • Suitable for general-purpose pipetting and high-throughput workflows.
Catalog No. 50-273-3210