Learn More
Bellco Glass, Inc. 1000µL Pipette Tips, LTS Fit, Sterile
SDP
Supplier:  Bellco Glass, Inc. 229076000
Encompass_Preferred
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tipsPipette tips are packaged in racks with hinged lids designed for easy one-handed opening10μL extended length tips have a maximum volume of 20μL and are compatible with 2, 10, and 20μL pipettesFine tip for improved accuracy and precisionFilter tips are fitted with a polymer barrier to reduce the risk of contamination and prevent aerosols from penetrating into the pipetteLids feature stacking supports, and are removable to save bench space Pack Description96 Tips/RackCase Quantity960SterileYesRNase/DNase FreeYesAutoclavableYesLow RetentionYesFilterNoRackedYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume1000µLGraduatedYes
Catalog No. 50-232-9432