Learn More
Pacer Global LLC 1000μL Extra Long LTS-Compatible Pipette Tips, Filtered, Sterile, Low Retention, 96 tips/rack, 10 racks/pack.
SDP
Supplier:  Pacer Global LLC MLSPTFSLTS1000/DEL
Encompass_Preferred
1000μL Extra Long LTS-Compatible Pipette Tips, Filtered, Sterile, Low Retention, 96 tips/rack, 10 racks/pack. Lot traceable and DNase free, RNase free, and non-pyrogenic. Additional info on MLS-PTFS-LTS1000: Made of ultra-clear, medical-grade virgin polypropylene. Molded volume graduation markings. Ultra-low ejection force for comfort, ergonomic pipetting for delivering superior accuracy. Lids have a secure locking tab. Packaged in blue rigid autoclavable racks. Filters reduce cross contamination and prevent damaging aerosols from penetrating into the pipette. Racks and grids made from up to 61% recycled materials. MADE IN THE USA. Certifications: ISO13485:2016& ISO9001:2015 These pipette tips are LTS-Compatible and are compatible with Rainin single- and multi-channel LiteTouchpipettors.
Catalog No. NC2609844