Learn More
Sino Biological Recombinant Human basic FGF/FGF2 Protein
Supplier:  Sino Biological 10014HNAE10
Encompass_Preferred
A DNA sequence encoding the mature form of human bFGF (NP_001997.5) (Pro143-Ser288) was expressed with an additional Met at the N-terminus.
Catalog No. 50-161-3807