Learn More
Sino Biological Anti-Cytokeratin 7/KRT7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100310T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human KRT7 / Cytokeratin-7, and purified by antigen affinity chromatography.
Catalog No. 50-161-0852