Learn More
Sino Biological Anti-IFITM1/CD225 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100417T44100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human CD225, and purified by antigen affinity chromatography.
Catalog No. 50-161-1121