Learn More
Sino Biological Recombinant Human BAFF/BLyS/TNFSF13B Protein (ECD)
Supplier:  Sino Biological 10056HNCH100
Encompass_Preferred
A DNA sequence encoding the soluble form of human BAFF (Q9Y275-1) (Ala 134-Leu 285) was expressed and purified.
Catalog No. 50-161-3903