Learn More
Medchemexpress LLC HY-12702 10mg , 10058-F4 CAS:403811-55-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1270210MG
Encompass_Preferred
HY-12702 10mg 10058-F4 CAS: 403811-55-2 10058-F4 is a c-Myc inhibitor that prevents c-Myc-Max dimerization and transactivation of c-Myc target gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9332