Learn More
Selleck Chemical LLC 10058-F4 25mg 403811-55-2
SDP
Supplier:  Selleck Chemical LLC S715325MG
Encompass_Preferred
10058-F4 is a c-Myc inhibitor that specificallly inhibits the c-Myc-Max interaction and prevents transactivation of c-Myc target gene expression. 10058-F4 promotes a caspase-3-dependent apoptosis and modulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0743143